Order Entry
Mexico
ContactUsLinkComponent
Somso® Striated Muscle Fiber Model
Somso® Striated Muscle Fiber Model
Catalog # 470029-600
Supplier:  MARCUS SOMMER
undefined
Somso® Striated Muscle Fiber Model
Catalog # 470029-600
Supplier:  MARCUS SOMMER
Supplier Number:  BS 36

Specifications

  • Description:
    Striated Muscle Fiber
  • Height:
    21 cm
  • Length:
    18 cm
  • No. of Keyed Items:
    19
  • Cat. no.:
    470029-600
  • Width:
    18 cm
  • Supplier No.:
    BS 36
  • No. of Parts:
    1

Specifications

About this item

This model was recreated from the latest electron microscopy interpretation.

  • Muscle Fibers from Two Sectional Planes
  • Motor Neuron Attachment Shows Innervation
  • Five-Year Manufacturer's Warranty

The accurate, high-quality model shows the innervation of muscle cells, multi-nuclear striated muscle fibers, medullated nerve fiber with Schwann cells and synaptic vessels, and the striated nature of myofibrils including Z lines and Q stripes.

The muscle fiber, enlarged 4000x, displays exceptional detail.