Order Entry
Mexico
ContactUsLinkComponent
 
Somso® Comprehensive Functional Larynx Model
  470029-638
undefined
Somso® Comprehensive Functional Larynx Model
  470029-638
 :  GS 10

 

  • Description:
    Comprehensive Functional Larynx
  • Height:
    33 cm
  • Length:
    18 cm
  • No. of Keyed Items:
    16
  • Cat. no.:
    470029-638
  • Width:
    18 cm
  • Supplier No.:
    GS 10
  • No. of Parts:
    1

 

 

Demonstrates Vocal Cord Movement

  • Vocal Cords Can Be Manipulated
  • Large Size For Easier Manipulation And Examination
  • 5 Year Manufacturers Warranty

Show how the larynx works with this 3X-enlarged, complete model featuring all structural components, including the hyoid bone, surrounding cartilage, and musculature. Students can re-create the motion of the larynx by increasing the tension on the drawstring to rotate the arytenoid cartilage, opening the vocal cords, making it perfect for lessons on speech, physiology, or anatomy.