Order Entry
Mexico
Orders LinkContactUsLinkComponent
Somso® Chromosome Model

Somso® Chromosome Model

Chromosome, 1, 18 cm, 18 cm, 47 cm
Catalog # 470029-434
Supplier:   MARCUS SOMMER
Supplier #:  ZOS 57/4
Somso® Chromosome Model
Catalog # 470029-434
Supplier:  MARCUS SOMMER
Somso® Chromosome Model
Somso® Chromosome Model

Chromosome, 1, 18 cm, 18 cm, 47 cm

Price
$14,550.00
Log in for availability
Each (1UN)
Product Details & Documents

Display A Metaphase Chromosome At 10,000X Magnification

  • Shows The Complex Structure Of Chromosomes
  • Removable From Base For Closer Examination
  • 5 Year Manufacturers Warranty

This unique model shows the kinetochores oriented to the outside of the chromosome, as well as the slightly irregular surface of the chromatids. Also shown are the chromatin loops coiled on the inside of the chromosome and the supported or imposed proteins. The lower section of the chromatid is exposed to reveal the chromatin.

Documents

Specifications
  • Description:
    Chromosome
  • Height:
    47 cm
  • Length:
    18 cm
  • Width:
    18 cm
  • No. of Parts:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.