Order Entry
Mexico
Orders LinkContactUsLinkComponent
Somso® Brain Ventricles Model

Somso® Brain Ventricles Model

Brain Ventricles, 1, 12 cm, 12 cm, 15 cm
Catalog # 470029-736
Supplier:   MARCUS SOMMER
Supplier #:  BS 24
Somso® Brain Ventricles Model
Catalog # 470029-736
Supplier:  MARCUS SOMMER
Somso® Brain Ventricles Model
Somso® Brain Ventricles Model

Brain Ventricles, 1, 12 cm, 12 cm, 15 cm

Price
$3,280.00
Log in for availability
Each (1UN)
Product Details & Documents

Enhances Understanding Of Brain Structures

  • Visualize The Ventricle Structure Not Visible In Brain Models
  • 5 Year Manufacturers Warranty

An excellent supplement to any full brain model, this model features only the four ventricles of the brain, making it ideal for in-depth study and relation to the rest of the brain’s structure. The model is mounted on a stand.

Documents

Specifications
  • Description:
    Brain Ventricles
  • Height:
    15 cm
  • Length:
    12 cm
  • Width:
    12 cm
  • No. of Parts:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.