Order Entry
Mexico
Orders LinkContactUsLinkComponent
Somso® Bony Labyrinth with Three Auditory Ossicles

Somso® Bony Labyrinth with Three Auditory Ossicles

Bony Labyrinth with Three Auditory Ossicles, 1, 12 cm, 12 cm, 3 cm
Catalog # 470030-006
Supplier:   MARCUS SOMMER
Supplier #:  QS 70/1
Somso® Bony Labyrinth with Three Auditory Ossicles
Catalog # 470030-006
Supplier:  MARCUS SOMMER
Somso® Bony Labyrinth with Three Auditory Ossicles
Somso® Bony Labyrinth with Three Auditory Ossicles

Bony Labyrinth with Three Auditory Ossicles, 1, 12 cm, 12 cm, 3 cm

Price
$4,050.00
Log in for availability
Each (1UN)
Product Details & Documents

Highly Accurate Replica Details The Complexity Of The Inner Ear

  • All Three Ossicles Cast In Proper Orientation With Bony Labyrinth
  • Enclosed In The Base For Protection From Loss Or Damage
  • 5 Year Manufacturers Warranty

The delicate structures of the bony structures of the inner ear are all replicated together. The malleus, incus, stapes, and bony labyrinth are detailed, mounted on a plastic base, and protectively enclosed in a Plexi-glass dome.

Documents

Specifications
  • Description:
    Bony Labyrinth with Three Auditory Ossicles
  • Height:
    3 cm
  • Length:
    12 cm
  • Width:
    12 cm
  • No. of Parts:
    1