Solderless Breadboard
Solderless Breadboard
Catalog # 470101-274
Supplier: JA ASSOCIATES
Supplier #: JAM JE21
Jump to:
Product Details & Documents
This solderless breadboard is ideal for high-frequency and low-noise circuits.
Perform your electronic experiments on this tiny breadboard (8×5.5×1 cm) . Interconnect any components with 20-29 AWG (3 to 8 mm) wire.
Its highly durable nickel-plated contacts provide a long life and low resistance connections. An added feature is the adhesive backing, allowing you to secure it anywhere.
This breadboard has a specially designed spring clip that’s made to withstand over five thousand insertion cycles.
Specifications
- Description:Solderless Breadboard