Order Entry
Mexico
ContactUsLinkComponent
Solar Heating Lab Activity
  470003-326
 :  Avantor
undefined
Solar Heating Lab Activity
  470003-326
 :  Avantor
  • Description:
    Solar Heating Lab Activity
  • No. of Lab Groups:
    1
  • Cat. no.:
    470003-326
  • Supplier No.:
    WARD470003-326
Working Solar Collector That Students Construct and Monitor.
  • Linked to a real world scenario
  • Allows direct observation of water circulation
  • Includes a teacher's guide
A basic, economical introduction to solar energy, this activity allows students to observe the simple gravity system used to circulate water and test the effects of different variables on the collector’s efficiency. As they do this, they will learn about the commercial applications of solar energy and even determine whether solar heating would be a viable option for their own homes. It includes a teacher’s guide.