Smooth Muscle Slide
Smooth Muscle Slide, Section, Hematoxylin and Eosin
Catalog # 470182-980
Supplier: Avantor
Jump to:
Product Details & Documents
Observe involuntary muscle characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human uterine wall.
Specifications
- Description:Smooth Muscle Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section
- Cat. No.:470182-980