Order Entry
Mexico
ContactUsLinkComponent
Skeletal Muscle Slide
Skeletal Muscle Slide
Catalog # 470182-678
Supplier:  Avantor
undefined
Skeletal Muscle Slide
Catalog # 470182-678
Supplier:  Avantor
Specifications
  • Description:
    Skeletal Muscle Slide
  • Cat. no.:
    470182-678
  • Stain:
    Hematoxylin and Eosin
  • Supplier No.:
    WARD470182-678
  • Orientation:
    Cross section
Specifications
About this item
Illustrates a voluntary muscle type
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Entire small muscle.