Order Entry
Mexico
ContactUsLinkComponent
Separating a Mixture by Physical Properties Kit
Separating a Mixture by Physical Properties Kit
Catalog # 470039-664
Supplier:  Avantor
undefined
Separating a Mixture by Physical Properties Kit
Catalog # 470039-664
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Separating a Mixture by Physical Properties Kit
  • No. of Lab Groups:
    10
  • Cat. no.:
    470039-664
  • Supplier No.:
    WARD470039-664
Specifications
About this item
Physically Separate Different Substances
  • Two Levels of Instruction Included
  • Determine properties of different substances and collect data
Students identify physical properties of substances and then determine methods that can separate a mixture made of those substances. Methods include separation by properties including particle size, density, solubility, and magnetism. Directions for two levels of instruction are included. The elementary level activity stresses identification properties, use of tools, and using collected data appropriately in designing an experiment. The advanced level activity includes some mathematical analysis for understanding physical properties of matter. one 45-minute class period is required to complete the activity. Includes Instruction manual. A balance is required but not included.
Includes:
Magnetic wands
Strainers
Funnels
Aluminum pie plates
Filter Paper
Plastic Spoons
Pop beads
Salt
Sand
Steel nuts
Pebbles
Plastic Cups
Plastic Containers
Instructions
Required but Not Provided: Balance, Beakers, Water, Containers, Paper Towels.