Scientific Method Problem Solving Kit
Scientific Method Problem-Solving Kit, 15
Catalog # 470002-462
Supplier: LAB-AIDS INC (KITS) SE
Supplier #: 100-A
Jump to:
Product Details & Documents
Easy-To-Understand, Practical Application Of Solving A Problem
- Materials For Up To 30 Students
- Form And Test A Hypothesis Using A Chemical Reaction
- Teachers Guide Included
Using observation and analysis of color changes in solutions with bromothymol blue indicator, students will employ the scientific method of defining a problem. They will collect data, formulate a hypothesis, and test the hypothesis, using both experimental and control data to conclude that a color change is usually evidence of a chemical reaction.
Documents
Specifications
- Description:Scientific Method Problem-Solving Kit
- No. of Lab Groups:15