Order Entry
Mexico
ContactUsLinkComponent
Salmonella enteritidis Slide
Salmonella enteritidis Slide
  470180-916
 :  Avantor
undefined
Salmonella enteritidis Slide
  470180-916
 :  Avantor
  • Description:
    Salmonella enteritidis
  • Cat. no.:
    470180-916
  • Stain:
    Gram (-)
  • Supplier No.:
    WARD470180-916
  • Orientation:
    Smear
Facultative Anerobe
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rods. Human intestinal pathogen.