Sable Brushes
Sable Brush, Sable hair, 5/16'', 3/32", 12", #4
470020-430
: 1014 SIZE4
Natural Brush Bristles, Appropriate for Many Specimens.
- Quality slide making supplies
- Soft and durable bristles for many uses
Use these brushes to position specimens on slides or into embedding material. They are fine-quality, red sable artist brushes with fine points that are extremely soft and pliable. They also feature rustproof, seamless aluminum ferrules with a solvent resistant setting.
- Bristle Material:Sable hair
- Brush head Ø:3/32"
- Description:Sable Brush
- Size:#4
- Total length:12"
- Brush Head Length:5/16''