Willow Branch Root (Salix species)
Catalog # 470177-474
Supplier: Avantor
Specifications
- Description:Branch Root of Willow
- Cat. no.:470177-474
- Stain:Quadruple
- Supplier No.:WARD470177-474
- Orientation:Cross Section
Specifications
About this item
Illustrates initial branch root growth.
- Identifying features clearly distinguishable
- Rigorous quality control standards
This slide demonstrates the beginning of the growth of a branch root. The branch root grows off of the major root of the plant.