Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rockwool Cubes
Rockwoold Cubes
  470145-010
 :  RWA03640
Rockwool Cubes
  470145-010
Rockwool Cubes
Rockwool Cubes
Rockwoold Cubes
$565.38
Kit de 98 Case de 30 (98UN)
Recommended for starting seedlings and cuttings.
  • Size: 1.5" x 1.5" x 1.5"
  • Sheet of 98
  • Description:
    Rockwoold Cubes