Rock Tumbler
Product Details & Documents
Encourage your students to look beyond the plain old rock they see everywhere with this rock tumbler. Your students will uncover the luster of rocks in 50% less time than more expensive units do. Use our heavy-duty rock tumbler to remove dirt and other undesirables from slabs of quartz, turquoise, and other rocks.
- Polish 1.78 kg of material at a time
- Mounted on a hardwood base
In fact, this versatile tumbler can polish, smooth, grind, cut, descale, and deburr rocks. You can even study the rocks during the various stages of the tumbling process. Durable tumbler is constructed of tough synthetic rubber and can be removed for easy cleaning. Both the motor and tumbler barrel are mounted on a 5.5x10" hardwood base.
Delivery information: Comes complete with #200 and #400 tumbling grit, polishing medium, adhesive pads and instructions. Powered with 115 VAC, it’s supplied with a 3-wire grounded cord and plug and includes instructions.
Specifications
- Description:Rock Tumbler