Order Entry
Mexico
Orders LinkContactUsLinkComponent
Retina Slide
Retina Slide
Retina Slide, Cross section, Hematoxylin and Eosin
Catalog # 470177-608
Supplier:   Avantor
Retina Slide
Catalog # 470177-608
Supplier:  Avantor
Retina Slide
Retina Slide
Retina Slide, Cross section, Hematoxylin and Eosin
Price
$193.60
Log in for availability
Each (1UN)
Product Details & Documents
Explore structures of the eye
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Shows retina, choroid, and sclera.
Specifications
  • Description:
    Retina Slide
  • Stain:
    Hematoxylin and Eosin
  • Orientation:
    Cross section