Resonance: The Velocity of Sound Apparatus
Catalog # 470101-098
Supplier: Avantor
Specifications
- Description:Resonance: The Velocity of Sound Apparatus
- Cat. no.:470101-098
- Supplier No.:WARD470101-098
Specifications
About this item
In this kit, ½ wavelengths are measured and used to determine the relationship between wavelength, frequency, and velocity.
- Developed by physics teacher, Jerome Chandler
- Grades 9 to 12
- Length: 122 cm
Simply fill tubes with water, strike a tuning fork above the inside tube, and adjust the inside tube to locate the ½ wavelength nodes. This easy-to-use kit helps students understand the properties of sound waves. Instructions included.
Ordering information: Tuning forks are required but not included.