Resistance Coils
Resistance Coils, Set/8
470005-710
A set of eight coils wound on individual plastic spools. Each coil has two binding posts.
A wooden pegboard base (not included) may be used to support the coils, labeled as follows:
Silver Nickel Wire AWG 30 Length 200 cm
Silver Nickel Wire AWG 30 Length 200 cm
Silver Nickel Wire AWG 30 Length 150 cm
Silver Nickel Wire AWG 30 Length 120 cm
Silver Nickel Wire AWG 30 Length 80 cm
Silver Nickel Wire AWG 30 Length 40 cm
Silver Nickel Wire AWG 28 Length 200 cm
Copper Wire AWG 30 Length 2000 cm
- Description:Resistance Coils, Set/8