Radiometer
Radiometer
470148-644
This simple apparatus demonstrates radiant heat.
- Diameter: 3"
- Height: 5"
- Vanes Turn When Dark Sides Absorb Heat
Vanes on the rotating shaft are light on one side but dark on the other.
When the dark sides of the vanes absorb light, the vanes turn.
- Description:Radiometer