Order Entry
Mexico
ContactUsLinkComponent
PYREX® Low-Form Beakers with Pour-Outs
PYREX® Low-Form Beakers with Pour-Outs
Catalog # 470211-266
Supplier:  CORNING CA
undefined
PYREX® Low-Form Beakers with Pour-Outs
Catalog # 470211-266
Supplier:  CORNING CA
Supplier Number:  1010

Specifications

  • Capacity:
    3000 ml (101 oz.)
  • Description:
    Pyrex® Low-Form Beakers with Pour-outs, With Handle and Stainless Steel Band
  • Cat. no.:
    470211-266
  • Supplier No.:
    1010

Specifications

About this item

Made of PYREX® borosilicate glass.

  • Excellent chemical resistance
  • Good chemical resistance

A sturdy, molded glass handle is secured to this 3 L beaker by a stainless steel strap which fits into a recess in the beaker wall. Hot liquids can be handled easily and safely.