Order Entry
Mexico
Orders LinkContactUsLinkComponent
PYREX® Measuring Pipettes

PYREX® Measuring Pipettes

2 ml, Pyrex® measuring pipettes, 0.1 ml
Catalog # 470211-586
Supplier:   CORNING CA
Supplier #:  7065-2
PYREX® Measuring Pipettes
Catalog # 470211-586
Supplier:  CORNING CA
PYREX® Measuring Pipettes
PYREX® Measuring Pipettes

2 ml, Pyrex® measuring pipettes, 0.1 ml

Price
$476.00
Log in for availability
Each (1UN) Case de 12 (1UN)
Product Details & Documents

Calibrated to deliver, these Mohr-type pipettes are color-coded by size for easy identification.

  • Colored graduations are enameled onto glass
  • Calibrated 'to deliver' (TD) their total capacity without blow-out

Pipette is reusable with round tip.

Documents

Specifications
  • Capacity:
    2 ml
  • Description:
    Pyrex® measuring pipettes
  • Subdivisions:
    0.1 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.