Order Entry
Mexico
Orders LinkContactUsLinkComponent
PYREX® Crystallizing Dishes

PYREX® Crystallizing Dishes

Crystallizing Dish, Pyrex, 100 mm, 50 mm
Catalog # 470211-452
Supplier:   CORNING CA
Supplier #:  3140-100
PYREX® Crystallizing Dishes
Catalog # 470211-452
Supplier:  CORNING CA
PYREX® Crystallizing Dishes
PYREX® Crystallizing Dishes

Crystallizing Dish, Pyrex, 100 mm, 50 mm

Price
$753.00
Log in for availability
Each (1UN) Case de 18 (1UN)
Product Details & Documents

PYREX® borosilicate glass, without spout.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.

Documents

Specifications
  • Ø:
    100 mm
  • Description:
    Crystallizing Dish, Pyrex
  • Height:
    50 mm