Pulley Bench Clamp Fitting
Pulley Bench Clamp Fitting
Catalog # 470120-950
Supplier: VWR International
Jump to:
Product Details & Documents
A Steady Hold
- Designed for clamping to the edge of a bench, table, and more, this pulley overhangs the edge in a vertical plane.
The device consists of a 50mm diameter pulley carried on a two part aluminum frame. Maximum clamp opening is 25mm. Overall frame length is 110mm.
Specifications
- Description:Pulley Bench Clamp Fitting