Pollution and the Environment Chart
Pollution and the Environment Chart, 38", 27"
470023-606
: 3345-L
Show Pollution Sources and Effects
Illustrate how various pollutants affect atmospheric changes, global warming, and marine life. The poster also shows changes to the environment that have occurred as a result of forest fires, atmospheric gases, greenhouse effects, and glacial melting, and then links these occurrences to animal extinction, solar radiation, and thermal balances. Laminated. Size: 27" x 38".
- Width:38"
- Description:Pollution and the Environment Chart
- Height:27"



