Pocket Stereoscope
Pocket Stereoscope
Catalog # 470021-864
Supplier: PECO SALES INC SE
Supplier #: 51154
Jump to:
Product Details & Documents
Glass Lenses Mean Greater Durability
- Size: 5 x 23/4" folded
This stereoscope not only folds to a convenient carrying size and comes with its own pocket case, it also features an adjustable interpupillary distance from 50 – 75 mm (2 – 3") for easy viewing.
The glass lenses are accurately ground for clear images with sharp definition.
Specifications
- Magnification:2.25×
- Description:Pocket Stereoscope
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



