Pneumatic Polypropylene Trough
Pneumatic Polypropylene Trough
Catalog # 470157-212
Supplier: Thermo Fisher Scientific
Supplier #: 6820-0010
Jump to:
Product Details & Documents
Use this timesaver for virtually any gas-collecting need. Setup is fast and easy with a built-in bridge that won't get lost, so the gas supply inlet can't be flooded. It is constructed of impact-resistant polypropylene that is lightweight and virtually unbreakable. It holds four 8 oz. bottles (not included); and the inlet connection and overflow outlet take ¼" Int.Ø tubing (not included).
- Size: 12¼L×6¾W×3"D
Specifications
- Description:Pneumatic Polypropylene Trough