Order Entry
Mexico
ContactUsLinkComponent
Plastic Slide and Coverslip Lab Pack
Plastic Slide and Coverslip Lab Pack
  470148-952
undefined
Plastic Slide and Coverslip Lab Pack
  470148-952
 :  20205104
  • Description:
    Plastic slides and coverslip lab pack
  • Cat. no.:
    470148-952
  • Supplier No.:
    20205104
Start making your own slides!
  • Explore slidemaking with this starter pack
  • Hands-on learning
  • Coverslips are made of clear PETG (polyethylene terephthalate glycol)
The set includes 50 slides 1×3", 0.5 mm thick and fifty 22 mm square coverslips and cleaning tissue.