Order Entry
Mexico
Orders LinkContactUsLinkComponent
Plastic Protractor
Plastic Protractor
Plastic Protractor
Catalog # 470151-092
Supplier:   ACME UNITED CORP SE
Supplier #:  376
Plastic Protractor
Catalog # 470151-092
Plastic Protractor
Plastic Protractor
Plastic Protractor
Price
$54.81
Log in for availability
Each (1UN) Paquete de 12 (1UN) Case de 144 (1UN)
Product Details & Documents
Made of transparent plastic with scale printed on one side. Graduated 0 to 180° and 180 to 0° in 0.5° divisions. Diameter: 6".
Specifications
  • Description:
    Plastic Protractor