Order Entry
Mexico
ContactUsLinkComponent
Ward's® Plasmolysis in Plant Cells Lab Activity
Ward's® Plasmolysis in Plant Cells Lab Activity
  470015-332
 :  Avantor
undefined
Ward's® Plasmolysis in Plant Cells Lab Activity
  470015-332
 :  Avantor
  • Description:
    WARD’S Plasmolysis in Plant Cells Lab Activity
  • Cat. no.:
    470015-332
  • Supplier No.:
    WARD470015-332
Observe Dramatic Effect of Water Loss on Onion Cells.
  • Aligned to NGSS standards (MS/HS: LS1)
What makes a plant wilt when it needs water, and “come back to life” when it receives that water? Find out with this simple investigation. Using only an onion you supply and this kit’s materials, students can easily see the effect of varying osmotic conditions on plant cells, and learn the importance of water to a plant’s structure. It includes enough materials for six setups, a teacher’s guide, and student copymaster.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.