Order Entry
Mexico
ContactUsLinkComponent
Plasmodium falciparum, Ring Stages Slide
Plasmodium falciparum, Ring Stages Slide
Catalog # 470181-948
Supplier:  Avantor
undefined
Plasmodium falciparum, Ring Stages Slide
Catalog # 470181-948
Supplier:  Avantor
Specifications
  • Description:
    Plasmodium falciparum, Ring Stages Slide
  • Cat. no.:
    470181-948
  • Stain:
    Giemsa
  • Supplier No.:
    WARD470181-948
  • Orientation:
    Smear
Specifications
About this item
Human blood parasite
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
From human blood. Agent of malignant tertian Malaria.
To learn more about this organism, please visit the CDC's article about the stages of Plasmodium falciparum in blood: https://www.cdc.gov/dpdx/resources/pdf/benchAids/malaria/Pfalciparum_benchaidV2.pdf