Permanent Ceramic Disc Magnet
Permanent Ceramic Disc Magnet
470105-152
: CD022800
A permanent ceramic disc magnet with the North and South poles on opposite ends of the diameter.
58 mm diameter and 6.4 mm thick.
- Description:Permanent Ceramic Disc Magnet