Order Entry
Mexico
Orders LinkContactUsLinkComponent
Pendulum Clamp
Pendulum Clamp
Pendulum Clamp
Certificates
Pendulum Clamp
1 Options Available Below
Product Details & Documents
This sturdy clamp is invaluable for experiments involving energy, motion, acceleration, and more.
  • Clamp Hangs Three Pendulums at Once
  • Easy-To-Adjust Pendulum Lengths
  • Length: 10⅝"
  • Topics Include Energy, Motion, Acceleration, and More
It hangs three pendulums at once, and you can adjust any pendulum without disturbing the others. The support arm screws into the included rod clamp and has thumbscrews for suspension cords.
10⅝" in length; it can be used with a 1/2" or 3/4" dia. vertical support rod.
Product Family Options
Product Information
  • Availability
    Price
  • 470006-526
    Pendulum Clamp, 1 Pc.
    Pendulum Clamp, 1 Pc.
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $300.60
    $300.60
    Each (1UN)
       $300.60
    Supplier Number: CLPEN1
  • Discontinued
    Supplier Number: -