Peat Pots
Peat Pots, 2 1/4"
Catalog # 470147-452
Supplier: PARK SEED WHOLESALE
Supplier #: 00624-CS-SP
Jump to:
Product Details & Documents
Composed of peat moss and wood fiber, these pots allow the seedling to be planted without removing the pot. They allow easy penetration by growing roots and decompose as the plant develops.
: Package of 24.
Specifications
- Description:Peat Pots, 2 1/4"