Order Entry
Mexico
ContactUsLinkComponent
Paramecium multimicronucleatum Slide
Paramecium multimicronucleatum Slide
  470181-474
 :  Avantor
undefined
Paramecium multimicronucleatum Slide
  470181-474
 :  Avantor
  • Description:
    Paramecium multimicronucleatum Slide
  • Cat. no.:
    470181-474
  • Stain:
    Schneider's
  • Supplier No.:
    WARD470181-474
  • Orientation:
    Explano-Mount™
Easy to see internal structures
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Larger than P. caudatum with a large macronucleus. Micronuclei are variable in number and arranged in a row.