Order Entry
Mexico
ContactUsLinkComponent
Paramecium caudatum, Explano-Mount Slide
Paramecium caudatum, Explano-Mount Slide
  470183-646
 :  Avantor
undefined
Paramecium caudatum, Explano-Mount Slide
  470183-646
 :  Avantor
  • Description:
    Paramecium caudatum, Explano-Mount
  • Cat. no.:
    470183-646
  • Stain:
    Fast Green and Mallory's
  • Supplier No.:
    WARD470183-646
  • Orientation:
    Explano-Mount™
Slide and Illustrated Guide Packed in a Durable Plastic Sleeve
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Shows general structures.