Order Entry
Mexico
Orders LinkContactUsLinkComponent
Parafilm Dispenser

Parafilm Dispenser

Parafilm Dispenser
Catalog # 470202-830
Supplier #:  HS234525B
Parafilm Dispenser
Catalog # 470202-830
Parafilm Dispenser
Parafilm Dispenser

Parafilm Dispenser

Price
$1,378.37
Log in for availability
Each (1UN) Case de 30 (1UN)
Product Details & Documents

For maximum protection, this dispenser is designed with a safety razor that slides and cuts with no danger to fingers.

  • ABS Plastic Construction
  • Dispenser Holds One or Two 2" Rolls or one 4" Roll
  • Blue
  • Safety Razor Included

The dispenser holds one or two 2" rolls or one 4" roll.

The dispenser is made of blue ABS plastic.

Specifications
  • Description:
    Parafilm Dispenser
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.