Palatine Tonsil Slide
Catalog # 470183-020
Supplier: Avantor
Specifications
- Description:Palatine Tonsil Slide
- Cat. no.:470183-020
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470183-020
- Orientation:Section
Specifications
About this item
Located in the back of the human throat.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human.