Order Entry
Mexico
ContactUsLinkComponent
 
Owl Pellet Dissection Chart Set
  470012-454
 :  Avantor
undefined
Owl Pellet Dissection Chart Set
  470012-454
 :  Avantor

 

  • Width:
    43 cm
  • Description:
    Owl Pellet Charts
  • Length:
    58 cm
  • Cat. no.:
    470012-454
  • Finish:
    Unlaminated
  • Supplier No.:
    21

 

 

Detailed Visual Guides For Classroom Experiments

  • Set of eight simple and effective charts

Charts lend understanding to the characteristics of owls as well as provide guidance in the owl pellet dissection process. Each chart includes vivid illustrations and instructive information. Charts include the owl’s food web, a bone sorting chart, a pellet information chart, and skeletal diagrams of a shrew, bird, mole and vole.