Olfactory Epithelium Slide
Olfactory Epithelium, Thin Section, Hematoxylin and Eosin
470183-672
: Avantor
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows pseudostratified columnar epithelium with no goblet cells and no distinct basal lamina.
- Description:Olfactory Epithelium
- Stain:Hematoxylin and Eosin
- Orientation:Thin Section