Hooked Masses, Set of 9
470200-626
- Description:Hooked Mass Set, Set of 9
- Cat. no.:470200-626
- Supplier No.:56206-01S
- No. of Weights:9 (10N, 5N, Two 2N, 1N, 0.5N, Two 0.2N, 0.1N)
Hooked Mass Sets provide accurate results.
- Stainless Steel Construction
- Accurate Results
- Lathe-Turned for Maximum Precision
- Plastic Storage Block Included
These hook weights are made of stainless steel.
The weights are lathe turned for maximum precision.
: This set includes a plastic storage block.
: One set includes nine weights.