Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Oceanic Circulation Model
Ward's® Oceanic Circulation Model
Ward's® Oceanic Circulation Model
Catalog # 470015-808
Supplier #:  80-0255
Ward's® Oceanic Circulation Model
Catalog # 470015-808
Ward's® Oceanic Circulation Model
Ward's® Oceanic Circulation Model
Ward's® Oceanic Circulation Model
Price
$3,484.80
Log in for availability
Each (1UN)
Product Details & Documents
Examine Oceanic Water Patterns That Cause the Gulf Stream and El Niño.
  • Complex ocean water circulation patterns become clear with our easy-to-observe working model
Run warm water, representing equatorial water, and cold 'polar' water together to re-create density currents that circulate vertically.
It comes with two clear plastic tubes, two clear plastic connecting tubes, two clear plastic containers, food coloring, salt, and a user’s guide.
Tube size: Ø 1½ × 12" L
Connecting tube size: Ø ½×8¼" L
Container size: 3⅞" cube.
Detailed instruction manual with suggested activities provided.
Specifications
  • Description:
    Ward's® Oceanic Circulation Model