About this item
Designed for today's crowded laboratory benchtops!
- Compact footprint is over 40% smaller than other benchtop meters
- Large display permits easier readout viewing
- Datalogging allows you to store and recall up to 100 data sets
The easy-to-use, microprocessor-based pH 700 benchtop meter measures pH, mV, relative mV, and temperature. The large, dual-display LCD provides excellent visibility—even from a distance. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures. Five-point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.
Meter features HOLD, ATC, selectable °F or °C measurement, recall of electrode slope and offset, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 470019-672
- 470019-670
- pH range:
- ‒2 to 16
- pH resolution:
- 0.01/0.1
- pH accuracy:
- ±0.01 pH
- Automatic buffer recognition:
- NIST and USA
- mV range:
- ‒2000 to 2000
- mV resolution:
- 0.1 or 1 mV
- mV accuracy:
- ±0.2 mV or 0.05%, whichever is greater
- Temperature range:
- 0...100 °C (32...212 °F)
- Display:
- LCD
- Power supply:
- 100 - 240 VAC, 50/60 Hz
- Dimensions:
- Height: 2.75"; Width: 6.875"
- Interfaces:
- None
- Calibration:
- Up to 6 points