Morchella, Asci Slide
Catalog # 470181-066
Supplier: Avantor
Specifications
- Description:Morchella, Asci
- Cat. no.:470181-066
- Stain:Quadruple
- Supplier No.:WARD470181-066
- Orientation:Section
Specifications
About this item
Mushroom Spore Distributing Structure
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mature fruiting body of morel.