Order Entry
Mexico
ContactUsLinkComponent
Mixed Ciliates Slide
  470181-462
 :  Avantor
undefined
Mixed Ciliates Slide
  470181-462
 :  Avantor
  • Description:
    Mixed Ciliates
  • Cat. no.:
    470181-462
  • Stain:
    Fast Green and Mallory's
  • Supplier No.:
    WARD470181-462
  • Orientation:
    Explano-Mount™
Protozoans Usually Found in Water and Soil
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Variety of common species are shown.