Ward's® Mitosis And Meiosis Manipulative Model
Catalog # 470003-986
Supplier: Avantor
Specifications
- Description:Mitosis And Meiosis Manipulative Model
- No. of Lab Groups:10
- Cat. no.:470003-986
- Supplier No.:WARD470003-986
Specifications
About this item
Study Cell Division With This Comprehensive Learning Activity.
- Pre- and post-laboratory assessments
- Links to real world concepts
- Estimated class/lab time required: 30-40 minutes
- Aligned to NGSS standards (MS/HS: LS1, LS3)
By using the various bases and chromosomes, students can manipulate the different stages that occur during mitosis and meiosis, and discover the various changes that take place within the cell. All model parts are laminated for durability. Partial assembly is required. Includes 10 sets with teacher’s guide and suggested activities. Grades 7–10.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.