Mini Sharpener
Mini Sharpener
Catalog # 470016-784
Supplier: LANSKY SHARPENERS
Supplier #: LCKEY
Jump to:
Product Details & Documents
Perfect for sharpening dull scalpels and dissecting needles.
This lightweight, compact sharpener uses two high-quality aluminum-ceramic rods mounted in a hard plastic base to restore sharpness and precision to the dissection equipment. Includes key chain for easy transport. Color may vary. Size: 2" W×2 3/8" H×3/4" D.
Specifications
- Description:Mini Sharpener