Order Entry
Mexico
Orders LinkContactUsLinkComponent
Microscale Thermometers

Microscale Thermometers

Microscale Thermometers, 0...100 °C
Catalog # 470018-442
Supplier #:  4-428
Microscale Thermometers
Catalog # 470018-442
Microscale Thermometers
Microscale Thermometers

Microscale Thermometers, 0...100 °C

Price
$1,012.70
Log in for availability
Paquete de 1
Product Details & Documents

Miniature thermometers (6 cm long) for use in microscale chemistry.

Bulbs have a 5 mm diameter and will fit the smallest microplate well. Packages of 12.

Specifications
  • Description:
    Microscale Thermometers
  • Range:
    0...100 °C