Order Entry
Mexico
ContactUsLinkComponent
Acrylic Micropipette Stand
Acrylic Micropipette Stand
Catalog # 470019-914
undefined
Acrylic Micropipette Stand
Catalog # 470019-914
Supplier Number:  HS206203

Specifications

  • Description:
    3-place micropipette stand
  • Cat. no.:
    470019-914
  • Supplier No.:
    HS206203

Specifications

About this item

Convenient and safe storage for pipettes.

  • Holds three pipettes
  • Designed to fit most major pipette brands
  • Made of clear acrylic

Upper slot: 28 mm
Lower slot: 20 mm