SP Bel-Art Mercury Collector, Bel-Art Products, a part of SP
Catalog # 470153-828
Supplier: BEL-ART PRODUCTS SE
Specifications
- Description:Mercury Collector
- Cat. no.:470153-828
- Supplier No.:F362500000
Specifications
About this item
Simply press the foam collector onto elusive beads of spilled mercury and the foam pores open to enclose it. Closing the container forces the beads into a recessed trap. One replacement pad is included.